Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 148531000
Price (USD)/ Each
88.65
Online exclusive
104.00 
Save 15.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
88.65
Online exclusive
104.00 
Save 15.35 (15%)
Add to cart
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Catalog number148531000
Price (USD)/ Each
88.65
Online exclusive
104.00 
Save 15.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,4,6-trimethyl-1,3,5-triazinane-1,3,5-triium
Molecular FormulaC6H18N3
InChI KeyMZSSRMMSFLVKPK-UHFFFAOYSA-Q
SMILESCC1[NH2+]C(C)[NH2+]C(C)[NH2+]1
Molecular Weight (g/mol)132.23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only