Hexachloroethane, 99%, Thermo Scientific Chemicals
Hexachloroethane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexachloroethane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 148541000
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
30.00 
Save 4.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
30.00 
Save 4.35 (15%)
Add to cart
Hexachloroethane, 99%, Thermo Scientific Chemicals
Catalog number148541000
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
30.00 
Save 4.35 (15%)
-
Add to cart
Chemical Identifiers
CAS67-72-1
IUPAC Namehexachloroethane
Molecular FormulaC2Cl6
InChI KeyVHHHONWQHHHLTI-UHFFFAOYSA-N
SMILESClC(Cl)(Cl)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Crystalline powder or crystals
Melting point185°C to 195°C
Infrared spectrumConforms
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only