Thermo Scientific Chemicals

Methyl dichloroacetate, 99%, Thermo Scientific Chemicals

Catalog number: 148621000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methyl dichloroacetate, 99%, Thermo Scientific Chemicals

Catalog number: 148621000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 148621000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
116-54-1
IUPAC Name
methyl 2,2-dichloroacetate
Molecular Formula
C3H4Cl2O2
InChI Key
HKMLRUAPIDAGIE-UHFFFAOYSA-N
SMILES
COC(=O)C(Cl)Cl
Appearance (Form)
Liquid
GC
>=98.5 %
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4411 to 1.4431 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text