1x1 image pixel for data collection
Methyl cyanoacetate, 97%
Methyl cyanoacetate, 97%
Methyl cyanoacetate, 97%
Methyl cyanoacetate, 97%
Thermo Scientific Chemicals

Methyl cyanoacetate, 97%

CAS: 105-34-0 | C4H5NO2 | 99.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number 148642500
Price (USD)
38.65
Special offer
Online exclusive
Ends: 26-Sep-2025
47.40
Save 8.75 (18%)
Each
In stock
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
38.65
Special offer
Online exclusive
Ends: 26-Sep-2025
47.40
Save 8.75 (18%)
Each
Add to cart
Chemical Identifiers
CAS105-34-0
IUPAC Namemethyl 2-cyanoacetate
Molecular FormulaC4H5NO2
InChI KeyANGDWNBGPBMQHW-UHFFFAOYSA-N
SMILESCOC(=O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Refractive index1.4170 to 1.4190 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection