Trimethyl orthoformate, 99%, Thermo Scientific Chemicals
Trimethyl orthoformate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethyl orthoformate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
500 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 148660050
Price (USD)/ Each
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS149-73-5
IUPAC Nametrimethoxymethane
Molecular FormulaC4H10O3
InChI KeyPYOKUURKVVELLB-UHFFFAOYSA-N
SMILESCOC(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3780 to 1.3800 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only