Thermo Scientific Chemicals

Cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 148671000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 148671000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 148671000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2043-61-0
IUPAC Name
cyclohexanecarbaldehyde
Molecular Formula
C7H12O
InChI Key
KVFDZFBHBWTVID-UHFFFAOYSA-N
SMILES
O=CC1CCCCC1
GC
>=96.0 %
Appearance (Color)
Clear colorless to light pink
Infrared spectrum
Conforms
Refractive index
1.4490 to 1.4520 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text