Thermo Scientific Chemicals

4-Bromobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 148761000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 148761000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 148761000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-58-8
IUPAC Name
4-bromobenzene-1-sulfonyl chloride
Molecular Formula
C6H4BrClO2S
InChI Key
KMMHZIBWCXYAAH-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1=CC=C(Br)C=C1
Appearance (Form)
Crystalline powder or crystals
Melting point
74°C to 77°C
Appearance (Color)
White to beige
Infrared spectrum
Conforms
Titration Argentometric
>=97.5 % (Ex CL)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text