Phenyl acetate, 97%, Thermo Scientific Chemicals
Phenyl acetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenyl acetate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number 148771000
Price (USD)/ Each
42.70
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
42.70
Add to cart
Phenyl acetate, 97%, Thermo Scientific Chemicals
Catalog number148771000
Price (USD)/ Each
42.70
-
Add to cart
Chemical Identifiers
IUPAC Namephenyl acetate
Molecular FormulaC8H8O2
InChI KeyIPBVNPXQWQGGJP-UHFFFAOYSA-N
SMILESCC(=O)OC1=CC=CC=C1
Molecular Weight (g/mol)136.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to yellow to light brown
Infrared spectrumConforms
Refractive index1.4995 to 1.5035 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only