Thermo Scientific Chemicals

Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 148890500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 148890500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 148890500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
367-51-1
IUPAC Name
sodium 2-sulfanylacetate
Molecular Formula
C2H3NaO2S
InChI Key
GNBVPFITFYNRCN-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)CS
Appearance (Color)
White to off-white to light pink
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration Iodimetric
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text