Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number 148892500
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
Add to cart
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Catalog number148892500
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS367-51-1
IUPAC Namesodium 2-sulfanylacetate
Molecular FormulaC2H3NaO2S
InChI KeyGNBVPFITFYNRCN-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to light pink
Appearance (Form)Powder
Infrared spectrumConforms
Titration Iodimetric>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only