Thermo Scientific Chemicals

3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: 148902500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: 148902500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 148902500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1119-62-6
IUPAC Name
3-[(2-carboxyethyl)disulfanyl]propanoic acid
Molecular Formula
C6H10O4S2
InChI Key
YCLSOMLVSHPPFV-UHFFFAOYSA-N
SMILES
OC(=O)CCSSCCC(O)=O
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
154°C to 158°C
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text