Thiamine hydrochloride, 98.5-101.5%
Thiamine hydrochloride, 98.5-101.5%
Thiamine hydrochloride, 98.5-101.5%
Thiamine hydrochloride, 98.5-101.5%
Thermo Scientific Chemicals

Thiamine hydrochloride, 98.5-101.5%

Thiamine hydrochloride, 98.5-101.5%, C12H17ClN4OS.HCl, CAS Number-67-03-8, aneurine hydrochloride, berin, thiamin chloride, thiamine chloride hydrochloride, thiamine dichloride, thiamine hcl, thiamine hydrochloride, thiaminium chloride, trophite, vitamin b1 hydrochloride, 1kg, 98.5 to 101.5% | CAS: 67-03-8 | C12H17ClN4OS·HCl | 337.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 148991000
Price (USD)
73.65
Special offer
Online exclusive
Ends: 26-Sep-2025
90.80
Save 17.15 (19%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
Ends: 26-Sep-2025
90.80
Save 17.15 (19%)
Each
Add to cart
Chemical Identifiers
CAS67-03-8
IUPAC Namehydrogen 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium dichloride
Molecular FormulaC12H18Cl2N4OS
InChI KeyDPJRMOMPQZCRJU-UHFFFAOYSA-M
SMILES[H+].[Cl-].[Cl-].CC1=C(CCO)SC=[N+]1CC1=CN=C(C)N=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO498.5 to 101.5 % (On anhydrous substance)
Water=<5 % (K.F.)
Heavy metals (as Pb)=<20 ppm
Sulfated ash=<0.1 %
Sulfate (SO4)=<300 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link