Thermo Scientific Chemicals

Saccharin, 98+%, Thermo Scientific Chemicals

Catalog number: 149001000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Saccharin, 98+%, Thermo Scientific Chemicals

Catalog number: 149001000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 149001000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81-07-2
IUPAC Name
2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
Molecular Formula
C7H5NO3S
InChI Key
CVHZOJJKTDOEJC-UHFFFAOYSA-N
SMILES
O=C1NS(=O)(=O)C2=CC=CC=C12
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Melting point
226°C to 230°C
Titration with NaOH
98.0 to 101.0 % (On dried substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text