Thermo Scientific Chemicals

Propionic acid, sodium salt, 99.0-100.5%, Thermo Scientific Chemicals

Catalog number: 149010010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Propionic acid, sodium salt, 99.0-100.5%, Thermo Scientific Chemicals

Catalog number: 149010010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 149010010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
137-40-6
IUPAC Name
sodium propanoate
Molecular Formula
C3H5NaO2
InChI Key
JXKPEJDQGNYQSM-UHFFFAOYSA-M
SMILES
[Na+].CCC([O-])=O
Appearance (Form)
Crystalline or granular powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text