2-Methyl-2-propanethiol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methyl-2-propanethiol, 99%, Thermo Scientific Chemicals
Have Questions?
Quantity:
50 mL
Packaging:
Glass bottle
Catalog number 149020500
Price (USD)/ Each
21.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
25.70 
Save 4.05 (16%)
-
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
21.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
25.70 
Save 4.05 (16%)
Add to cart
2-Methyl-2-propanethiol, 99%, Thermo Scientific Chemicals
Catalog number149020500
Price (USD)/ Each
21.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
25.70 
Save 4.05 (16%)
-
Add to cart
Chemical Identifiers
CAS75-66-1
IUPAC Name2-methylpropane-2-thiol
Molecular FormulaC4H10S
InChI KeyWMXCDAVJEZZYLT-UHFFFAOYSA-N
SMILESCC(C)(C)S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4220 to 1.4240 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only