Methyl Propionate, 99+%
Methyl Propionate, 99+%
Methyl Propionate, 99+%
Methyl Propionate, 99+%
Thermo Scientific Chemicals

Methyl Propionate, 99+%

CAS: 554-12-1 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 149050010
Price (USD)
167.65
Online Exclusive
186.00
Save 18.35 (10%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
167.65
Online Exclusive
186.00
Save 18.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS554-12-1
IUPAC Namemethyl propanoate
Molecular FormulaC4H8O2
InChI KeyRJUFJBKOKNCXHH-UHFFFAOYSA-N
SMILESCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3750 to 1.3790 (20°C, 589 nm)
Water=<0.3 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link