Thermo Scientific Chemicals

Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals

Catalog number: 149062500
250 g, Plastic Bottle, Each
Thermo Scientific Chemicals

Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals

Catalog number: 149062500
250 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 149062500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2582-30-1
IUPAC Name
N''-aminoguanidine; carbonic acid
Molecular Formula
C2H8N4O3
InChI Key
OTXHZHQQWQTQMW-UHFFFAOYSA-N
SMILES
OC(O)=O.NN=C(N)N
Infrared spectrum
Conforms
Titration with HClO4
98.0 to 102.0 %
Residue after ignition
=<0.3 %
Appearance (Color)
White to cream to light yellow
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text