5-Norbornene-2-carboxaldehyde, 95%, mixture of endo and exo, Thermo Scientific Chemicals
5-Norbornene-2-carboxaldehyde, 95%, mixture of endo and exo, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Norbornene-2-carboxaldehyde, 95%, mixture of endo and exo, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 149101000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5453-80-5
IUPAC Namebicyclo[2.2.1]hept-5-ene-2-carbaldehyde
Molecular FormulaC8H10O
InChI KeyAJIBZRIAUXVGQJ-UHFFFAOYNA-N
SMILESO=CC1CC2CC1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
Refractive index1.4870 to 1.4920 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only