Thermo Scientific Chemicals

Iodoform, 99+%, Thermo Scientific Chemicals

Catalog number: 149120025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Iodoform, 99+%, Thermo Scientific Chemicals

Catalog number: 149120025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 149120025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-47-8
IUPAC Name
triiodomethane
Molecular Formula
CHI3
InChI Key
OKJPEAGHQZHRQV-UHFFFAOYSA-N
SMILES
IC(I)I
Appearance (Color)
Bright yellow to dark yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
119°C to 122°C
Loss on drying
=<0.5 % (24 h) (Over silicagel)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text