n-Amyl acetate, 99%
n-Amyl acetate, 99%
n-Amyl acetate, 99%
Thermo Scientific Chemicals

n-Amyl acetate, 99%

n-Amyl acetate, 99%, C7H14O2, CAS Number-628-63-7, 1-pentyl acetate, acetic acid, pentyl ester, amyl acetate, amyl acetic ester, amyl acetic ether, birnenoel, n-amyl acetate, n-pentyl acetate, n-pentyl ethanoate, pent-acetate, 2.5L, 98.5% min. (GC), 02, 131, 149.0 deg.C, 100.0, CHEBI:87362 | CAS: 628-63-7 | C7H14O2 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass bottle
Catalog number 149180025
Price (USD)
161.65
Special offer
Online exclusive
Ends: 26-Sep-2025
190.00
Save 28.35 (15%)
Each
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
161.65
Special offer
Online exclusive
Ends: 26-Sep-2025
190.00
Save 28.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS628-63-7
IUPAC Namepentyl acetate
Molecular FormulaC7H14O2
InChI KeyPGMYKACGEOXYJE-UHFFFAOYSA-N
SMILESCCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4010 to 1.4030 (20°C, 589 nm)
Appearance (Form)Liquid
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471915Certificate of AnalysisMay 17, 2025149180025, 149182500, 149180010
A0465629Certificate of AnalysisAug 22, 2024149180025, 149182500, 149180010
A0462054Certificate of AnalysisApr 17, 2024149180025, 149182500, 149180010
A0457743Certificate of AnalysisNov 14, 2023149180025, 149182500, 149180010
A0455000Certificate of AnalysisAug 25, 2023149180025, 149182500, 149180010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link