Thermo Scientific Chemicals

Sodium formaldehydesulfoxylate dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 149190250
25 kg, Fibre drum, Each
Thermo Scientific Chemicals

Sodium formaldehydesulfoxylate dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 149190250
25 kg, Fibre drum, Each
Quantity
Packaging
Catalog number: 149190250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6035-47-8
IUPAC Name
sodium dihydrate sulfinomethanolate
Molecular Formula
CH7NaO5S
InChI Key
VJWVWUHJLZEHOU-UHFFFAOYNA-N
SMILES
O.O.[Na+].OS(=O)C[O-]
Appearance (Color)
White
Zinc (Zn)
=<30 ppm
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration Iodometric
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text