Thermo Scientific Chemicals

Methacrylamide, 98%, extra pure

Catalog number: 149202500
250 g, Each
Thermo Scientific Chemicals

Methacrylamide, 98%, extra pure

Catalog number: 149202500
250 g, Each
Quantity
Catalog number: 149202500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-39-0
IUPAC Name
2-methylprop-2-enamide
Molecular Formula
C4H7NO
InChI Key
FQPSGWSUVKBHSU-UHFFFAOYSA-N
SMILES
CC(=C)C(N)=O
Water
=<1 % (K.F.)
Appearance (Color)
White to light yellow
Appearance (Form)
Crystals
Infrared spectrum
Conforms
Melting point
108°C to 112°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text