Methacrylamide, 98%, extra pure
Methacrylamide, 98%, extra pure
Thermo Scientific Chemicals

Methacrylamide, 98%, extra pure

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
2.5 kg
Catalog number 149205000
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.80 
Save 9.15 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.80 
Save 9.15 (15%)
Add to cart
Methacrylamide, 98%, extra pure
Catalog number149205000
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.80 
Save 9.15 (15%)
-
Add to cart
Chemical Identifiers
CAS79-39-0
IUPAC Name2-methylprop-2-enamide
Molecular FormulaC4H7NO
InChI KeyFQPSGWSUVKBHSU-UHFFFAOYSA-N
SMILESCC(=C)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 % (K.F.)
Appearance (Color)White to light yellow
Appearance (Form)Crystals
Infrared spectrumConforms
Melting point108°C to 112°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only