1-Hydroxy-2-naphthoic acid, 98%, Thermo Scientific Chemicals
1-Hydroxy-2-naphthoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Hydroxy-2-naphthoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
View all
Plastic bottle
Catalog number 149231000
Price (USD)/ Each
61.20
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
61.20
Add to cart
1-Hydroxy-2-naphthoic acid, 98%, Thermo Scientific Chemicals
Catalog number149231000
Price (USD)/ Each
61.20
-
Add to cart
Chemical Identifiers
CAS86-48-6
IUPAC Name1-hydroxynaphthalene-2-carboxylic acid
Molecular FormulaC11H8O3
InChI KeySJJCQDRGABAVBB-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C2C=CC=CC2=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to beige to grey-brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Melting point195°C to 205°C (Dec)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only