Thermo Scientific Chemicals

N-Chlorosuccinimide, 97%, Thermo Scientific Chemicals

Catalog number: 149250025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

N-Chlorosuccinimide, 97%, Thermo Scientific Chemicals

Catalog number: 149250025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 149250025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
128-09-6
IUPAC Name
1-chloropyrrolidine-2,5-dione
Molecular Formula
C4H4ClNO2
InChI Key
JRNVZBWKYDBUCA-UHFFFAOYSA-N
SMILES
ClN1C(=O)CCC1=O
Solubility
(5 % in acetone) Clear to light hazy colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text