Propionic acid, 99%, pure, Thermo Scientific Chemicals
Propionic acid, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Propionic acid, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Plastic drum
Catalog number 149300100
Price (USD)/ Each
162.65
Online exclusive
180.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
162.65
Online exclusive
180.00 
Save 17.35 (10%)
Add to cart
Propionic acid, 99%, pure, Thermo Scientific Chemicals
Catalog number149300100
Price (USD)/ Each
162.65
Online exclusive
180.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS79-09-4
IUPAC Namepropanoic acid
Molecular FormulaC3H6O2
InChI KeyXBDQKXXYIPTUBI-UHFFFAOYSA-N
SMILESCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Infrared spectrumConforms
Water=<0.2 %
Refractive index1.3850 to 1.3870 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only