Propylamine, 98%, pure, Thermo Scientific Chemicals
Propylamine, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Propylamine, 98%, pure, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number 149310010
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
96.60 
Save 13.95 (14%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
96.60 
Save 13.95 (14%)
Add to cart
Propylamine, 98%, pure, Thermo Scientific Chemicals
Catalog number149310010
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
96.60 
Save 13.95 (14%)
-
Add to cart
Chemical Identifiers
CAS107-10-8
IUPAC Namepropan-1-amine
Molecular FormulaC3H9N
InChI KeyWGYKZJWCGVVSQN-UHFFFAOYSA-N
SMILESCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only