Isopropanol, 99.5+%, pure, Thermo Scientific Chemicals
Isopropanol, 99.5+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropanol, 99.5+%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
5 L
25 L
Packaging:
View all
Plastic Drum
Catalog number 149320100
Price (USD)/ Each
284.65
Online exclusive
316.00 
Save 31.35 (10%)
-
Add to cart
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)/ Each
284.65
Online exclusive
316.00 
Save 31.35 (10%)
Add to cart
Isopropanol, 99.5+%, pure, Thermo Scientific Chemicals
Catalog number149320100
Price (USD)/ Each
284.65
Online exclusive
316.00 
Save 31.35 (10%)
-
Add to cart
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Infrared spectrumConforms
GC>=99.50 %
Water=<0.2 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only