Thermo Scientific Chemicals

2-Picoline, 98%, Thermo Scientific Chemicals

Catalog number: 149330025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

2-Picoline, 98%, Thermo Scientific Chemicals

Catalog number: 149330025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 149330025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
109-06-8
IUPAC Name
2-methylpyridine
Molecular Formula
C6H7N
InChI Key
BSKHPKMHTQYZBB-UHFFFAOYSA-N
SMILES
CC1=CC=CC=N1
Infrared spectrum
Conforms
GC
>=97.5 %
Water
=<0.2 %
Refractive index
1.4990 to 1.5010 (20°C, 589 nm)
Appearance (Color)
Clear light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text