Phenol, 99%, extra pure
Phenol, 99%, extra pure
Phenol, 99%, extra pure
Phenol, 99%, extra pure
Thermo Scientific Chemicals

Phenol, 99%, extra pure

CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 kg
50 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 149340010
Price (USD)
135.65
Special offer
Online exclusive
Ends: 26-Sep-2025
167.00
Save 31.35 (19%)
Each
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
135.65
Special offer
Online exclusive
Ends: 26-Sep-2025
167.00
Save 31.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS108-95-2
IUPAC Namephenol
Molecular FormulaC6H6O
InChI KeyISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point39°C to 42°C
Appearance (Color)Colorless to white
Appearance (Form)Crystals or fused mass
Infrared spectrumConforms
Water=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link