4-Methyl-2-pentanol, 99+%, Thermo Scientific Chemicals
4-Methyl-2-pentanol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methyl-2-pentanol, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
5 mL
500 mL
1 L
2.5 L
Packaging:
View all
Plastic drum
Catalog number 149380100
Price (USD)/ Each
-
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-methylpentan-2-ol
Molecular FormulaC6H14O
InChI KeyWVYWICLMDOOCFB-UHFFFAOYNA-N
SMILESCC(C)CC(C)O
Molecular Weight (g/mol)102.18
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4100 to 1.4120 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only