3-Hydroxypropionitrile, 99%, ACROS Organics™
3-Hydroxypropionitrile, 99%, ACROS Organics™
Thermo Scientific Chemicals

3-Hydroxypropionitrile, 99%, ACROS Organics™

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number 149392500
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
Add to cart
3-Hydroxypropionitrile, 99%, ACROS Organics™
Catalog number149392500
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name3-hydroxypropanenitrile
Molecular FormulaC3H5NO
InChI KeyWSGYTJNNHPZFKR-UHFFFAOYSA-N
SMILESOCCC#N
Molecular Weight (g/mol)71.08
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Refractive index1.4240 to 1.4280 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only