2,6-Dimethyl-4-heptanone, technical, remainder mainly 4,6-dimethyl-2-heptanone, Thermo Scientific Chemicals
2,6-Dimethyl-4-heptanone, technical, remainder mainly 4,6-dimethyl-2-heptanone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dimethyl-4-heptanone, technical, remainder mainly 4,6-dimethyl-2-heptanone, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
20 L
1 L
2.5 L
Packaging:
View all
Metal drum
Catalog number 149440200
Price (USD)/ Each
442.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
520.00 
Save 77.35 (15%)
-
Add to cart
Quantity:
20 L
Packaging:
Metal drum
Request bulk or custom format
Price (USD)/ Each
442.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
520.00 
Save 77.35 (15%)
Add to cart
2,6-Dimethyl-4-heptanone, technical, remainder mainly 4,6-dimethyl-2-heptanone, Thermo Scientific Chemicals
Catalog number149440200
Price (USD)/ Each
442.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
520.00 
Save 77.35 (15%)
-
Add to cart
Chemical Identifiers
CAS108-83-8
IUPAC Name2,6-dimethylheptan-4-one
Molecular FormulaC9H18O
InChI KeyPTTPXKJBFFKCEK-UHFFFAOYSA-N
SMILESCC(C)CC(=O)CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=90.0 % (main compound + 4.6-Dimethyl-2-heptanone)
GC10 to 30 % (4.6-Dimethyl-2-heptanone)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only