Diisopropylamine, 99%, Thermo Scientific Chemicals
Diisopropylamine, 99%, Thermo Scientific Chemicals
Diisopropylamine, 99%, Thermo Scientific Chemicals
Diisopropylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diisopropylamine, 99%, Thermo Scientific Chemicals

Diisopropylamine, 99%, C6H15N, CAS Number-108-18-9, 2-propanamine, n-1-methylethyl, bis propan-2-yl amine, diisopropylamine, di-isopropylamine, diisopropyl amine, dipa, n-1-methylethyl-2-propanamine, n-isopropyl-1-amino-2-methylethane, n-isopropylpropan-2-amine, n,n-diisopropylamine, 1L, 04, 154
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 149460025
Price (EUR)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS108-18-9
IUPAC Namebis(propan-2-yl)amine
Molecular FormulaC6H15N
InChI KeyUAOMVDZJSHZZME-UHFFFAOYSA-N
SMILESCC(C)NC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3910 to 1.3930 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 2
Acute oral toxicity
Category 4
Acute Inhalation Toxicity - Vapors
Category 3
Skin Corrosion/Irritation
Category 1
Serious Eye Damage/Eye Irritation
Category 1
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Danger

Hazard Statements

H302 - Harmful if swallowed

H314 - Causes severe skin burns and eye damage

H331 - Toxic if inhaled

H335 - May cause respiratory irritation

Physical Hazards

H225 - Highly flammable liquid and vapor

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P304 + P340 - IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Share catalog number, name or link