Ethyl acetate, 99+%, extra pure
Ethyl acetate, 99+%, extra pure
Ethyl acetate, 99+%, extra pure
Ethyl acetate, 99+%, extra pure
Thermo Scientific Chemicals

Ethyl acetate, 99+%, extra pure

CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 149470025
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS141-78-6
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Water=<0.05 % (K.F.)
Residue after evaporation=<0.005 %
Appearance (Form)Clear liquid
Color scale=<10 APHA
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link