Ethyl acetate, 99+%, Thermo Scientific Chemicals
Ethyl acetate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl acetate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
5 L
10 L
Packaging:
View all
Metal Drum
Catalog number 149470250
Price (USD)/ Each
583.65
Online exclusive
686.00 
Save 102.35 (15%)
-
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (USD)/ Each
583.65
Online exclusive
686.00 
Save 102.35 (15%)
Add to cart
Ethyl acetate, 99+%, Thermo Scientific Chemicals
Catalog number149470250
Price (USD)/ Each
583.65
Online exclusive
686.00 
Save 102.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
Molecular Weight (g/mol)88.11
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Water=<0.05 % (K.F.)
Residue after evaporation=<0.005 %
Appearance (Form)Clear liquid
Color scale=<10 APHA
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only