1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 149480025
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
Add to cart
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
Catalog number149480025
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS71-23-8
IUPAC Namepropan-1-ol
Molecular FormulaC3H8O
InChI KeyBDERNNFJNOPAEC-UHFFFAOYSA-N
SMILESCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Residue after evaporation=<0.005 %
Refractive index1.3840 to 1.3860 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only