Thermo Scientific Chemicals

Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 149520050
5 L, Plastic Drum, Each
Thermo Scientific Chemicals

Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 149520050
5 L, Plastic Drum, Each
Quantity
Packaging
Catalog number: 149520050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-05-8
IUPAC Name
acetonitrile
Molecular Formula
C2H3N
InChI Key
WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES
CC#N
Appearance (Form)
Clear liquid
Water
=<0.3 % (K.F.)
Residue after evaporation
=<0.005 %
Color scale
=<20 APHA
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text