Valeraldehyde, 97%, Thermo Scientific Chemicals
Valeraldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Valeraldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 149561000
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
25.90 
Save 3.25 (13%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
25.90 
Save 3.25 (13%)
Add to cart
Valeraldehyde, 97%, Thermo Scientific Chemicals
Catalog number149561000
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
25.90 
Save 3.25 (13%)
-
Add to cart
Chemical Identifiers
CAS110-62-3
IUPAC Namepentanal
Molecular FormulaC5H10O
InChI KeyHGBOYTHUEUWSSQ-UHFFFAOYSA-N
SMILESCCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3925 to 1.3975 (20°C, 589 nm)
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only