Valeraldehyde, 97%, Thermo Scientific Chemicals
Valeraldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Valeraldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
1 L
Packaging:
Glass bottle
Catalog number 149562500
Price (USD)/ Each
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepentanal
Molecular FormulaC5H10O
InChI KeyHGBOYTHUEUWSSQ-UHFFFAOYSA-N
SMILESCCCCC=O
Molecular Weight (g/mol)86.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3925 to 1.3975 (20°C, 589 nm)
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only