Thermo Scientific Chemicals

Valeric Acid, 99%

Catalog number: 149570010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Valeric Acid, 99%

Catalog number: 149570010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 149570010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-52-4
IUPAC Name
pentanoic acid
Molecular Formula
C5H10O2
InChI Key
NQPDZGIKBAWPEJ-UHFFFAOYSA-N
SMILES
CCCCC(O)=O
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Water
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text