1x1 image pixel for data collection
Ethanolamine, 99%
Ethanolamine, 99%
Ethanolamine, 99%
Thermo Scientific Chemicals

Ethanolamine, 99%

CAS: 141-43-5 | C2H7NO | 61.08 g/mol
Have Questions?
Catalog NumberQuantity
149582500250 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 149582500
Price (USD)
33.65
Online Exclusive
37.00
Save 3.35 (9%)
Each
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)
33.65
Online Exclusive
37.00
Save 3.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS141-43-5
IUPAC Name2-aminoethan-1-ol
Molecular FormulaC2H7NO
InChI KeyHZAXFHJVJLSVMW-UHFFFAOYSA-N
SMILESNCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<15 APHA
Infrared spectrumConforms
GC>=98.5 %
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link