Tetraethylene glycol, 99.5%, Thermo Scientific Chemicals
Tetraethylene glycol, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetraethylene glycol, 99.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 149590010
Price (USD)/ Each
56.65
Online exclusive
65.90 
Save 9.25 (14%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
56.65
Online exclusive
65.90 
Save 9.25 (14%)
Add to cart
Tetraethylene glycol, 99.5%, Thermo Scientific Chemicals
Catalog number149590010
Price (USD)/ Each
56.65
Online exclusive
65.90 
Save 9.25 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-{2-[2-(2-hydroxyethoxy)ethoxy]ethoxy}ethan-1-ol
Molecular FormulaC8H18O5
InChI KeyUWHCKJMYHZGTIT-UHFFFAOYSA-N
SMILESOCCOCCOCCOCCO
Molecular Weight (g/mol)194.23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4581 to 1.4600 (20°C, 589 nm)
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only