Acetic acid, sodium salt, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, sodium salt, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 149610025
Price (USD)/ Each
181.65
Online exclusive
202.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
181.65
Online exclusive
202.00 
Save 20.35 (10%)
Add to cart
Acetic acid, sodium salt, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals
Catalog number149610025
Price (USD)/ Each
181.65
Online exclusive
202.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS127-09-3
IUPAC Namesodium acetate
Molecular FormulaC2H3NaO2
InChI KeyVMHLLURERBWHNL-UHFFFAOYSA-M
SMILES[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<20 ppm
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO4>=99.0 %
Loss on drying=<1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only