(±)-Propylene oxide, 99%, pure
(±)-Propylene oxide, 99%, pure
(±)-Propylene oxide, 99%, pure
(±)-Propylene oxide, 99%, pure
Thermo Scientific Chemicals

(±)-Propylene oxide, 99%, pure

(+/-)-Propylene oxide, 99%, C3H6O, CAS Number-75-56-9, 2,3-epoxypropane, epoxypropane, methyloxirane, propylene oxide, oxirane, methyl, propyleneoxide, 1,2-propylene oxide, propene oxide, propylene epoxide, 1,2-epoxypropane, 1L, 17, 6, 34.0 deg.C, CHEBI:38685, Undesignated, 0.8300g/mL, 200-879-2 | CAS: 75-56-9 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 149620010
Price (USD)
53.65
Special offer
Online exclusive
Ends: 26-Sep-2025
66.30
Save 12.65 (19%)
Each
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 26-Sep-2025
66.30
Save 12.65 (19%)
Each
Add to cart
Chemical Identifiers
CAS75-56-9
IUPAC Name2-methyloxirane
Molecular FormulaC3H6O
InChI KeyGOOHAUXETOMSMM-UHFFFAOYNA-N
SMILESCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3650 to 1.3670 (20°C, 589 nm)
Aldehyde (C3H6O)=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link