Thermo Scientific Chemicals

(±)-Propylene oxide, 99%, pure, Thermo Scientific Chemicals

Catalog number: 149620025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

(±)-Propylene oxide, 99%, pure, Thermo Scientific Chemicals

Catalog number: 149620025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 149620025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-56-9
IUPAC Name
2-methyloxirane
Molecular Formula
C3H6O
InChI Key
GOOHAUXETOMSMM-UHFFFAOYNA-N
SMILES
CC1CO1
Refractive index
1.3650 to 1.3670 (20°C, 589 nm)
Aldehyde (C3H6O)
=<0.01 %
Appearance (Form)
Clear liquid
Water
=<0.02 %
Color scale
=<10 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text