Acrylonitrile, 99+%
Acrylonitrile, 99+%
Acrylonitrile, 99+%
Thermo Scientific Chemicals

Acrylonitrile, 99+%

CAS: 107-13-1 | C3H3N | 53.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 149630010
Price (EUR)
43,61
Online Exclusive
51,30
Save 7,69 (15%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
43,61
Online Exclusive
51,30
Save 7,69 (15%)
Each
Add to cart
Chemical Identifiers
CAS107-13-1
IUPAC Nameprop-2-enenitrile
Molecular FormulaC3H3N
InChI KeyNLHHRLWOUZZQLW-UHFFFAOYSA-N
SMILESC=CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Stabilizer35 to 45 ppm MEHQ
Water=<0.5 %
Infrared spectrumConforms
Color scale=<25 APHA
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472303Certificate of AnalysisMay 17, 2025149630100, 149630025, 149631000, 149630010
A0469559Certificate of AnalysisFeb 05, 2025149630100, 149630025, 149631000, 149630010
A0469560Certificate of AnalysisFeb 04, 2025149630100, 149630025, 149631000, 149630010
A0463137Certificate of AnalysisMay 23, 2024149630100, 149630025, 149630010, 149631000
A0449386Certificate of AnalysisJan 22, 2024149630100, 149630025, 149630010, 149631000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 2
Acute oral toxicity
Category 3
Acute dermal toxicity
Category 3
Acute Inhalation Toxicity - Vapors
Category 3
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 1
Skin Sensitization
Category 1
Carcinogenicity
Category 1B
Reproductive Toxicity
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Chronic aquatic toxicity
Category 2
Label Elements
Signal Word

Danger

Hazard Statements

H301 + H311 + H331 - Toxic if swallowed, in contact with skin or if inhaled

H315 - Causes skin irritation

H317 - May cause an allergic skin reaction

H318 - Causes serious eye damage

H335 - May cause respiratory irritation

H350 - May cause cancer

H361 - Suspected of damaging fertility or the unborn child

H411 - Toxic to aquatic life with long lasting effects

Physical Hazards

H225 - Highly flammable liquid and vapor

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Additional EU labelling

Restricted to professional users

Share catalog number, name or link

1x1 image pixel for data collection