Thermo Scientific Chemicals

Acrylonitrile, 99+%, Thermo Scientific Chemicals

Catalog number: 149630025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Acrylonitrile, 99+%, Thermo Scientific Chemicals

Catalog number: 149630025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 149630025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-13-1
IUPAC Name
prop-2-enenitrile
Molecular Formula
C3H3N
InChI Key
NLHHRLWOUZZQLW-UHFFFAOYSA-N
SMILES
C=CC#N
Appearance (Form)
Clear liquid
Water
=<0.5 %
Stabilizer
35 to 45 ppm MEHQ
Infrared spectrum
Conforms
Color scale
=<25 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text