Thermo Scientific Chemicals

Di-n-butyl ether, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 149690025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

Di-n-butyl ether, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 149690025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 149690025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142-96-1
IUPAC Name
1-butoxybutane
Molecular Formula
C8H18O
InChI Key
DURPTKYDGMDSBL-UHFFFAOYSA-N
SMILES
CCCCOCCCC
Appearance (Form)
Clear liquid
GC
>=99.0 %
Color scale
=<10 APHA
Infrared spectrum
Conforms
Water
=<0.05 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text