Di-n-butyl ether, 99+%, extra pure, Thermo Scientific Chemicals
Di-n-butyl ether, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-n-butyl ether, 99+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal drum
Catalog number 149690250
Price (USD)/ Each
-
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Chemical Identifiers
CAS142-96-1
IUPAC Name1-butoxybutane
Molecular FormulaC8H18O
InChI KeyDURPTKYDGMDSBL-UHFFFAOYSA-N
SMILESCCCCOCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.0 %
Color scale=<10 APHA
Infrared spectrumConforms
Water=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only